Learn More
MilliporeSigma™ Novagen™ Sf9 Insect Cells
GSA_VA
Catalog No. 711043 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
3 vials
Catalog No. Quantity
71-104-3 3 vials
1 options
Catalog No. 71-104-3 Supplier MilliporeSigma™ Supplier No. 711043
Add to Cart
Add to Cart
For establishment of cultures suitable for any application
  • Provided as frozen stocks of Spodoptera frugiperda Sf9 cells
  • Cells with BacVector™ Insect Cell Medium are recommended for cotransfection of transfer plasmids with BacVector Triple Cut Virus DNA
  • Cells with BacMagic™ DNA for construction of baculovirus recombinants and for transfection of pIEx7™ and pBiEx™ vector constructs of InsectDirect™System
  • After recovery, they can be grown as semi-adherent cultures in tissue culture flasks or in suspension as shaker cultures
  • Each vial contains 1x107 cells
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -140°C Freezer; Shipped on dry ice. Upon receipt, the vials should be removed from their container and either recovered immediately, or placed at -70°C and used within two weeks. For longer-term storage, place in liquid nitrogen.
Product Type Insect Cells
Quantity 3 vials