Learn More
Description
EMD Millipore Novagen™ SpinPrep™, Gel DNA Kit contains no organic extraction or alcohol precipitation.
- Extraction of DNA fragments of 150 bp to > 12,000 bp in size from agarose gels
- Uses GelMelt™ Solution to dissolve the gel slice
- Adsorption of DNA to silica membrane in spin column form
- Purified DNA is eluted in low-salt buffer
- Each spin column can bind up to 20 μg DNA
- Recoveries are consistently in range of 50 to 90% under standard conditions
- Packaging: Glass Bottle
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Format | Kit |
| Isolation Technology | Silica Membrane Technology |
| No. of Reactions | 100 Reactions/Kit |
| Product Type | DNA Extraction Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.