Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Streptavidin Agarose

Catalog No. 692033 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-203-3 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-203-3 Supplier MilliporeSigma™ Supplier No. 692033
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Consists of cross-linked agarose covalently coupled with pure streptavidin.

EMD Millipore Novagen™ Streptavidin Agarose minimizes leaching of streptavidin while maintaining full binding activity.

  • Used in column batch formats
  • Used with applications with high biotin binding capacity, low non-specific binding
  • No detectable protease, DNase, or RNase contamination
  • Specifications:
  • Protease None Detected (SDS-PAGE of purified protein)
  • DNase None Detected
  • RNase None Detected (gel assay of purified RNA)
  • Packaged in Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Quantity 5 mL
For Use With (Application) Column and batch formats
Type Agarose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.