Learn More
Description
- Directed against 11 amino acid gene 10 leader peptide expressed by many of the pET translation vectors
- Cross-reactivity: Negligible with bacterial, insect or mammalian cell lysates
- Sensitivity 1-10ng (Western blot developed with chromogenic substrates)
- Suitable for immunoprecipitation immunohistochemistry (1-3)
Specifications
Specifications
| Content And Storage | -20°C |
| Product Type | T7 Monoclonal Antibody |
| Quantity | 50 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.