Learn More
MilliporeSigma™ Novagen™ T7 Tail Fiber Monoclonal Antibody
Catalog No. 715303 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
71-530-3 50 μg
1 options
Catalog No. 71-530-3 Supplier MilliporeSigma™ Supplier No. 715303
Used to immunoprecipitate T7 phage or T7 tail-fiber from solution.
EMD Millipore Novagen™ T7 Tail Fiber Monoclonal Antibody is mouse monoclonal antibody(IgG2a) directed against tail-fiber protein of T7 bacteriophage.
  • Mouse monoclonal antibody
  • Uses an optimized immunoprecipitation GrabIt™ based protocol
  • 2.5μg of antibody is sufficient to immunoprecipitate at least 1 × 1010T7 phage
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 μg