Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ T7Select™, 1-1 Cloning Kit

Catalog No. 700103 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-010-3 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-010-3 Supplier MilliporeSigma™ Supplier No. 700103
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uses T7 capsid protein to display peptides or proteins on surface of phage.

EMD Millipore Novagen™, T7Select, 1-1 Cloning Kit decreases time needed to perform multiple rounds of growth.

  • Easy to use
  • Easy to grow replicates more rapidly
  • Plaques form within 3 hr at 37°C
  • Cultures lyse 1-2 hr after infection
  • Decreases time needed to perform multiple rounds of growth usually required for selection
  • Extremely robust, is stable to harsh conditions that inactivate other phage
  • Variety of agents that can be used in biopanning selection procedures
  • Easy to obtain in large amounts, efficient in vitro packaging system
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -70°C
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.