Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ T7Select™ 10-3 Cloning Kit

Catalog No. 705503 Shop All MilliporeSigma Products
Change view
Click to view available options
Content And Storage:
-70°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
70-550-3 -70°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-550-3 Supplier MilliporeSigma™ Supplier No. 705503
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uses T7 capsid protein to display peptides or proteins on surface of phage.

EMD Millipore Novagen™ T7Select™ 10-3 Cloning Kit is phage display system.

  • Phage display system
  • Displays 50 amino acids in size in high copy number (415 per phage), peptides or proteins up to about 1200 amino acids in low copy number (0.1-1 per phage) or mid-copy number (5-15 per phage)
  • Easy to grow replicates more rapidly
  • Plaques form within 3 hr at 37°C cultures lyse 1-2 hr after
  • Extremely robust
  • Stable to harsh conditions
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -70°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.