Learn More
MilliporeSigma™ Novagen™ T7Select™ Packaging 1-1 Cloning Kit
Catalog No. 700143 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
6 Ea./Kit
Catalog No. Quantity
70-014-3 6 Ea./Kit
1 options
Catalog No. 70-014-3 Supplier MilliporeSigma™ Supplier No. 700143
Add to Cart
Add to Cart
Includes
Six T7 Packaging Extracts, 1μg T7Select Packaging Control DNA, 0.2mL BL21 glycerol stock, 0.2mL BLT5403 glycerol stock, 0.2mL BLT5615 glycerol stock
Displays peptides up to about 50 amino acids in size.
EMD Millipore Novagen™ T7Select™ Packaging 1-1 Cloning Kit produces library containing at least 5 x 107 recombinants per microgram of arms.
  • Easy to use
  • High efficiency extracts (> 2 x 108 pfu/μg with T7Select Packaging Control DNA)
  • Extracts made from special engineered phage that reduces nonrecombinant cloning background to below 0.1%
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -70°C
Quantity 6 Ea./Kit