Learn More
MilliporeSigma™ Novagen™ Thrombin Cleavage Capture Kit
GSA_VA
Catalog No. 690223 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
69-022-3 1 Kit
1 options
Catalog No. 69-022-3 Supplier MilliporeSigma™ Supplier No. 690223
Add to Cart
Add to Cart
Designed for cleavage of fusion proteins.
EMD Millipore Novagen™ Thrombin Cleavage Capture Kit used with any fusion protein that contains a thrombin recognition sequence.
  • Convenient, quantitative removal of thrombin protease
  • Monitors performance of cleavage conditions
  • Easily visualized by SDS-PAGE
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Multiple storage requirements
Type Thrombin Cleavage Capture Kit
Quantity 1 Kit