Learn More
MilliporeSigma™ Novagen™ X-Gluc Solution
Catalog No. 700363 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
Catalog No. Quantity
70-036-3 500 μL
1 options
Catalog No. 70-036-3 Supplier MilliporeSigma™ Supplier No. 700363
Used to identify recombinants containing pBACgus vectors LIC GUS Control Insert.
EMD Millipore Novagen™ X-Gluc Solution is colorimetric stain for GUS activity.
  • Provided as 20mg/mL stock solution
  • Ready for dilution use in staining plaques for GUS activity
  • Each vial contains enough substrate to stain 15, 100mm plates
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
Quantity 500 μL
Storage Requirements −20°C