Learn More
MilliporeSigma™ Novagen™ YeastBuster™ Protein Extraction Reagent
Catalog No. 711864 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
71-186-4 Each
1 options
Catalog No. 71-186-4 Supplier MilliporeSigma™ Supplier No. 711864
Add to Cart
Add to Cart
Formulated for extraction of active proteins from yeast cells.
EMD Millipore Novagen™ YeastBuster™ Protein Extraction Reagent yields enzymatically active proteins.
  • Eliminates inconsistencies associated with abrasive grinding, ultrasonication, and pressure disruption of yeast cells
  • Fully compatible with Ni-NTA His·Bind™ and GST·Bind™ affinity purification methods
  • Avoids harsh conditions of vigorous mechanical or chemical treatment
  • Tested with Saccharomyces cerevisiae, Pichia pastoris, P.stipidis, and Schizosaccharomyces pombe strains and with plant cells
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each
Kit Contents NucBuster Extraction Reagent 1, 7.5mL NucBuster Extraction Reagent 2,100μ: 100mM DTT, Protease Inhibitor Cocktail Set I (Lyophilized, makes 100μL)
Content And Storage 2° to −8°C