Learn More
Medchemexpress LLC Novaluron | 116714-46-6 | 99.8% | 492.70 | 250 MG
SDP
Catalog No. 5000383911
Encompass_Preferred
Novaluron is a chemical with insecticide properties, an insect growth regulator, and has adverse effects on mouse sperm. For research use only, it is not sold to patients. Novaluron (0-100 μM, 90 min) inhibits the viability of mouse spermatocytes. Novaluron (10-100 μM, 90 min) significantly reduces the number of acrosome-reactive sperm, significantly reduces sperm motility, and changes PKA activity and tyrosine phosphorylation.
  • Insecticide properties
  • Insect growth regulator
  • Inhibits the viability of mouse spermatocytes
  • Reduces the number of acrosome-reactive sperm
  • Reduces sperm motility
  • Changes PKA activity and tyrosine phosphorylation
Catalog No. 50-003-83911 Supplier Medchemexpress LLC Supplier No. 250MGHY17519
Novaluron is a chemical with insecticide properties, an insect growth regulator, and has adverse effects on mouse sperm. For research use only, it is not sold to patients. Novaluron (0-100 μM, 90 min) inhibits the viability of mouse spermatocytes. Novaluron (10-100 μM, 90 min) significantly reduces the number of acrosome-reactive sperm, significantly reduces sperm motility, and changes PKA activity and tyrosine phosphorylation.
  • Insecticide properties
  • Insect growth regulator
  • Inhibits the viability of mouse spermatocytes
  • Reduces the number of acrosome-reactive sperm
  • Reduces sperm motility
  • Changes PKA activity and tyrosine phosphorylation