Learn More
AdipoGen Novaluron
SDP
Supplier:  AdipoGen CDXN0069M100
Encompass_Preferred
Chemical. CAS 116714-46-6. Formula C17H9ClF8N2O4. MW 492.7. Synthetic. Acylurea insecticide. Insect growth inhibitor. Chitin synthase inhibitor active against larvae of Coleoptera, Homoptera, Diptera and Lepidoptera. Shown to be an inhibitor of acetylcholinesterase in B-biotype Bemisia tabaci. Compound can be used as analytical reference material.
Catalog No. 50-256-0418