Learn More
BURKLE INC Novartos Multi, 23.62 in. (60 cm),double,8 chambers,witho inserts
Catalog No. 5000025580
Encompass_Preferred
With the Novartos Multi 2-compartment double sampler, a double multi-point sample can be taken from several predefined sampling zones. Depending on the length, four, seven or nine collection chambers are fitted with two sample inserts (accessories) each for the double sampler.
Catalog No. 50-000-25580 Supplier BURKLE INC Supplier No. 53580602
With the Novartos Multi 2-compartment double sampler, a double multi-point sample can be taken from several predefined sampling zones. Depending on the length, four, seven or nine collection chambers are fitted with two sample inserts (accessories) each for the double sampler.