Learn More
BURKLE INC Novartos Multi, 23.62 in. (60cm),triple,12 chambers,witho inserts
Catalog No. 5000025787
Encompass_Preferred
The Novartos Multi 3-fold sampler is designed for precise sampling according to the unit dose procedure, specifically tailored for the pharmaceutical industry. It collects a triple multipoint sample from multiple predefined sampling zones.
Depending on its length, the sampler features four, seven, or nine collection chambers. Please note that sample inserts are sold separately, allowing you to customize your sampling setup. Ideal for reliable and representative sampling of small powder portions in pharmaceutical quality control.
Catalog No. 50-000-25787 Supplier BURKLE INC Supplier No. 53580603
The Novartos Multi 3-fold sampler is designed for precise sampling according to the unit dose procedure, specifically tailored for the pharmaceutical industry. It collects a triple multipoint sample from multiple predefined sampling zones.
Depending on its length, the sampler features four, seven, or nine collection chambers. Please note that sample inserts are sold separately, allowing you to customize your sampling setup. Ideal for reliable and representative sampling of small powder portions in pharmaceutical quality control.