Learn More
BURKLE INC Novartos Multi, 39.37 in. (100cm),double,14 chambers,witho inser.
Catalog No. 5000025581
Encompass_Preferred
With the Novartos Multi 2-compartment double sampler, a double multi-point sample can be taken from several predefined sampling zones. Depending on the length, four, seven or nine collection chambers are fitted with two sample inserts (accessories) each for the double sampler.
Catalog No. 50-000-25581 Supplier BURKLE INC Supplier No. 53581000
With the Novartos Multi 2-compartment double sampler, a double multi-point sample can be taken from several predefined sampling zones. Depending on the length, four, seven or nine collection chambers are fitted with two sample inserts (accessories) each for the double sampler.