Learn More
BURKLE INC Novartos Multi, 55.12 in. (140 cm), triple, 27 chambers, witho inser.
Catalog No. 5000025584
Encompass_Preferred
The Novartos Multi 3-fold sampler is designed for precise sampling according to the unit dose procedure, specifically tailored for the pharmaceutical industry. It collects a triple multipoint sample from multiple predefined sampling zones.
Depending on its length, the sampler features four, seven, or nine collection chambers. Please note that sample inserts are sold separately, allowing you to customize your sampling setup. Ideal for reliable and representative sampling of small powder portions in pharmaceutical quality control.
Catalog No. 50-000-25584 Supplier BURKLE INC Supplier No. 53581403
The Novartos Multi 3-fold sampler is designed for precise sampling according to the unit dose procedure, specifically tailored for the pharmaceutical industry. It collects a triple multipoint sample from multiple predefined sampling zones.
Depending on its length, the sampler features four, seven, or nine collection chambers. Please note that sample inserts are sold separately, allowing you to customize your sampling setup. Ideal for reliable and representative sampling of small powder portions in pharmaceutical quality control.