Learn More
BURKLE INC Novartos Multi,55.12 in. (140 cm),double,18 chambers,witho inser.
Catalog No. 5000025569
Encompass_Preferred
With the Novartos Multi 2-compartment double sampler, a double multi-point sample can be taken from several predefined sampling zones. Depending on the length, four, seven or nine collection chambers are fitted with two sample inserts (accessories) each for the double sampler.
Catalog No. 50-000-25569 Supplier BURKLE INC Supplier No. 53581402
With the Novartos Multi 2-compartment double sampler, a double multi-point sample can be taken from several predefined sampling zones. Depending on the length, four, seven or nine collection chambers are fitted with two sample inserts (accessories) each for the double sampler.