Learn More
BURKLE INC Novartos Uno, 39.37 in. (100 cm), triple, 3 chambers,witho inser.
Catalog No. 5000026057
Encompass_Preferred
The Novartos Uno triple sampler takes a triple target point sample from a predefined sampling area. For the triple sampler, a collection chamber is equipped with three sample inserts (accessories).
Catalog No. 50-000-26057 Supplier BURKLE INC Supplier No. 53581030
The Novartos Uno triple sampler takes a triple target point sample from a predefined sampling area. For the triple sampler, a collection chamber is equipped with three sample inserts (accessories).