Learn More
BURKLE INC Novartos Uno, 23.62 in. (60 cm), double, 2 chambers,witho inserts
Catalog No. 5000026078
Encompass_Preferred
The Novartos Uno 2-compartment sampler is a high-precision unit dose sampler specifically designed for the pharmaceutical industry. Made from durable stainless steel, it enables double target point sampling from a predefined sampling area for maximum accuracy. The collection chamber is designed for use with two sample inserts (available separately) for double sampling. Perfect for pharmaceutical powder sampling and handling of very small quantities in GMP-compliant environments.
Catalog No. 50-000-26078 Supplier BURKLE INC Supplier No. 53580600
The Novartos Uno 2-compartment sampler is a high-precision unit dose sampler specifically designed for the pharmaceutical industry. Made from durable stainless steel, it enables double target point sampling from a predefined sampling area for maximum accuracy. The collection chamber is designed for use with two sample inserts (available separately) for double sampling. Perfect for pharmaceutical powder sampling and handling of very small quantities in GMP-compliant environments.