Learn More
BURKLE INC Novartos Uno, 23.62 in. (60 cm), triple, 3 chambers,witho inserts
Catalog No. 5000025673
Encompass_Preferred
The Novartos Uno triple sampler takes a triple target point sample from a predefined sampling area. For the triple sampler, a collection chamber is equipped with three sample inserts (accessories).
Catalog No. 50-000-25673 Supplier BURKLE INC Supplier No. 53580630
The Novartos Uno triple sampler takes a triple target point sample from a predefined sampling area. For the triple sampler, a collection chamber is equipped with three sample inserts (accessories).