Learn More
Ihc World LLC NOVAULTRA SIRIUS RED STAIN KIT
SDP
Catalog No. NC0249910
Encompass_Preferred
It's one of the best understood techniques and a sensitive method for collagen. The method is most frequently used on paraffin sections of tissues fixed adequately (at least 24 hours but ideally 1 or 2 weeks) in a neutral buffered formal
Catalog No. NC0249910 Supplier Ihc World LLC Supplier No. IW3012
It's one of the best understood techniques and a sensitive method for collagen. The method is most frequently used on paraffin sections of tissues fixed adequately (at least 24 hours but ideally 1 or 2 weeks) in a neutral buffered formal