Learn More
Ihc World LLC NOVAULTRA TOLUIDINE BLUE STAIN
SDP
Catalog No. NC0441447
Encompass
Mast cells are found in the connective tissue and their cytoplasm contains granules (metachromatic) composed of heparin and histamine. This kit is designed for staining of formalin fixed, paraffin embedded tissue sections. Mast cells will be stained
Catalog No. NC0441447 Supplier Ihc World LLC Supplier No. IW3013
Mast cells are found in the connective tissue and their cytoplasm contains granules (metachromatic) composed of heparin and histamine. This kit is designed for staining of formalin fixed, paraffin embedded tissue sections. Mast cells will be stained