Learn More
Sigma Aldrich Fine Chemicals Biosciences Novec 7000 Engineered Fluid, 375-03-1, MFCD01320779, 500 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SHH0001500ML
Encompass
This item has a minimum order quantity of 25 per supplier requirements. Linear Formula: CF3CF2CF2OCH3, Molecular Weight: 200.05, Synonym: 1-Methoxyheptafluoropropane, Methyl perfluoropropyl ether. Cryofreezing solution. ** For R&D use only. Not for drug, household, or other uses.
Catalog No. NC1104854