Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novec™ 7000 Engineered Fluid, Sigma-Aldrich

Catalog No. 01258981 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-981 500 mL
01-258-912 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-981 Supplier MilliporeSigma Supelco Supplier No. SHH0001500ML
Add to Cart
Add to Cart
  • Low Global Warming Potential/GWP (530, 100-year ITH)
  • Excellent dielectric properties: In event of leakage or other failure, will not damage electronic equipment
  • Zero ozone depletion potential (ODP)
  • Good materials compatibility
  • Low toxicity
  • Non-flammable
  • Non-corrosive
  • Good thermal stability
  • Useful at extreme low temperatures
  • Viscosity is less than 20 cSt at -120°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-03-1
Molecular Weight (g/mol) 250.06 g/mol

Specifications

Color Yellow
Quantity 500 mL
Packaging Amber Bottle
Physical Form Liquid
Percent Purity ≥99.5%
Chemical Name or Material Methoxy-nonafluorobutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.