Learn More
Sigma Aldrich Fine Chemicals Biosciences NovecTM 7100 Engneered fl500ML
Catalog No. 502900419
Encompass
Novec(TM) 7100 engineered fluid is a proprietary methoxy-nonafluorobutane from 3M(TM). This hydrofluoroether is a clear colorless and low-odor fluid intended to replace ozone-depleting substances (ODSs) and compounds with high global warming potential (GWP) in many applications. It has zero ozone depletion potential. High boiling point and low surface tension properties make it ideal for use in vapor degreasing applications as a neat (pure) azeotropic component or co-solvent parts cleaner. In addition its chemical and thermal stability non-flammability and low toxicity make it useful for many other industrial and specialty solvent applications.
Catalog No. 50-290-0419 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SHH0002500ML
Novec(TM) 7100 engineered fluid is a proprietary methoxy-nonafluorobutane from 3M(TM). This hydrofluoroether is a clear colorless and low-odor fluid intended to replace ozone-depleting substances (ODSs) and compounds with high global warming potential (GWP) in many applications. It has zero ozone depletion potential. High boiling point and low surface tension properties make it ideal for use in vapor degreasing applications as a neat (pure) azeotropic component or co-solvent parts cleaner. In addition its chemical and thermal stability non-flammability and low toxicity make it useful for many other industrial and specialty solvent applications.