Learn More
Medchemexpress LLC Novel bis-ortho-alkoxy-para-piperazinesubstituted-2,4-dianilinopyrimidines | 1472795-20-2 | 96.2% | 609.12 | C30H37ClN8O4 | 100MG
SDP
Catalog No. 5000239310
Encompass_Preferred
KRCA-0008 is a small-molecule dual inhibitor of anaplastic lymphoma kinase (ALK) and activated Cdc42 kinase 1 (Ack1/TNK2) intended for laboratory research use. It demonstrates potent enzymatic inhibition (ALK IC50 = 12 nM; Ack1 IC50 = 4 nM) and is supplied with high reported purity.
  • Potent dual inhibitor of ALK and Ack1 with IC50 values of 12 nM and 4 nM, respectively.
  • High purity (96.2%).
  • Molecular weight 609.12 and chemical formula C30H37ClN8O4.
  • Supplied in multiple pack sizes, including 100 mg.
  • Intended for preclinical and in vitro research use only; not for human or veterinary use.
Catalog No. 50-002-39310 Supplier Medchemexpress LLC Supplier No. HY12331100MG
KRCA-0008 is a small-molecule dual inhibitor of anaplastic lymphoma kinase (ALK) and activated Cdc42 kinase 1 (Ack1/TNK2) intended for laboratory research use. It demonstrates potent enzymatic inhibition (ALK IC50 = 12 nM; Ack1 IC50 = 4 nM) and is supplied with high reported purity.
  • Potent dual inhibitor of ALK and Ack1 with IC50 values of 12 nM and 4 nM, respectively.
  • High purity (96.2%).
  • Molecular weight 609.12 and chemical formula C30H37ClN8O4.
  • Supplied in multiple pack sizes, including 100 mg.
  • Intended for preclinical and in vitro research use only; not for human or veterinary use.