Learn More
Stoelting™ Novel Object Test
SDP
Catalog No. 10000352 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
manual
Catalog No. Model
10-000-352 manual
1 options
Catalog No. 10-000-352 Supplier Stoelting™ Supplier No. 62017
Add to Cart
Add to Cart
Stoelting™ Novel Object Test is based on the premise that rodents will explore a novel object more than a familiar one if the animal is able to remember the familiar one.
  • Following the training period, the rodent is removed from the environment for a delay period which can range from 5 minutes to 24 hours, depending on the type of memory being tested.
  • After the delay, the rodent is returned to the arena, where one of the original objects has been replaced by a new, dissimilar Novel Object.
  • The amount of time the rodent spends exploring each object provides a powerful measurement of memory integrity and attention.
Specifications
For Use With (Application) Neuroscience
Model manual