Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Novex™ 10%, Tris-Glycine, 1.0 mm, Midi Protein Gels, 12+2-well, w/adapters

Catalog No. WXP01012BXA
Encompass_Preferred
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WXP01012BXA 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. WXP01012BXA Supplier Invitrogen™ Supplier No. WXP01012BOXA
Save 22% on Invitrogen Gels, Reagents, and More
Use promo code GELS22
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Research Use Only. Not for use in diagnostic procedures.

Includes

Adapters for BioRad Criterion Cell

Separation Range: 30 to 250 kd, Separation Type: Denaturing, Native, For Use With: XCell4 SureLock Midi, BioRad Criterion

Novex™ Tris-Glycine Plus Midi Protein gels are improved polyacrylamide gels based ontraditional Laemmli protein electrophoresis, which allows the use of Laemmli sample and running buffers. Novex Tris-Glycine Plus gels offer reproducible separation of a wide range of proteins into well-resolved bands. The Novex Tris-Glycine Plus Midi Protein Gels are a 1.0 mm thick, wider format (8 cm x 13 cm) gel for higher throughput electrophoresis of protein samples. The included adapter is designed for use with the BioRad Criterion™ Cell.

  •  Improved shelf life—store gels for up to 12 months at 4°C
  • Diversity—use for native and denaturing protein assays
  • Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes

Recommended Storage

Store in refrigerator (2-8°C). Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

Gel Percentage 10%
Gel Size Midi
Gel Thickness 1.0 mm
Gel Type Tris-Glycine
Separation of Protein
Separation Range 30 to 250 kDa
Separation Type Denaturing, Native
Wells 12 + 2-well, w/adapters
Quantity 10/Pk.
For Use With (Equipment) Bio-Rad Criterion Cell
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.