Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Novex™ AP Chemiluminescent Substrate (CDP-Star™)

Catalog No. WP20002
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 100 ml bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WP20002 1 x 100 ml bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WP20002 Supplier Invitrogen™ Supplier No. WP20002
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Novex™ AP Chemiluminescent Substrate (CDP-Star™)

Novex™ AP Chemiluminescent Substrate is a non-radioactive ready-to-use solution of CDP-Star for chemiluminescence-based immunodetection of alkaline phosphatase (AP) on western blot or dot blot membranes. This reagent provides detection sensitivities superior to that of precipitating chromogenic substrates. Low picogram levels of detection can be achieved using either X-ray film or imaging equipment.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Single bottle labeled: Novex™ AP Chemiluminescent Substrate (CDP-Star™). Store at 4°C.
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Southern, Northern, and Western Blotting
Product Line Novex
Quantity 1 x 100 ml bottle
Substrate CDP-Star™
Substrate Properties Chemical Substrate
Substrate Type AP (Alkaline Phosphatase) Substrate
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.