Learn More
Invitrogen™ BenchMark™ His-tagged Protein Standard
Description
Includes
125μL (25 applications of 5μL each) is provided in loading buffer consisting of Tris-HCl, SDS, Glycerol, DTT, and Coomassie Blue G-250
The BenchMark His-tagged Protein Standard can be used with NuPAGE™ Gels and Tris-Glycine Gels.
- Verification of his-tagged fusion protein detection using InVision™ In-Gel Stain or an Anti-His(C-term) antibody
- Ten sharp and clear bands in the range of 10-160 kDa for molecular weight estimation of his-tagged proteins
- Compatibility with Coomassie™ R-250 stain
- A ready-to-use, time-saving format
- Gel Compatibility: NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels, SDS-PAGE Gels
1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Content And Storage | 125 μL (25 applications of 5 μL each) is provided in loading buffer consisting of Tris-HCl, SDS, Glycerol, DTT, and Coomassie™ Blue G-250. Store at -20°C. Avoid repeated freezing and thawing. |
| Detection Method | InVision™ His-tag In-gel Stain |
| Number of Markers | 10 |
| Ready to Load | Yes |
| Size Range | 10 to 160 kDa |
| Gel Compatibility | Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, SDS-PAGE Gels |
| Molecular Weight (g/mol) | 160, 120, 80, 60, 50, 40, 30, 20, 15, 10 kDa |
| Quantity | 125 μL |
| Shipping Condition | Dry Ice |
| Product Line | BenchMark |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.