Learn More
Invitrogen™ Large Gel Drying Kit
Catalog No. NI2207
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
NI2207 1 kit
1 options
Catalog No. NI2207 Supplier Invitrogen™ Supplier No. NI2207
Add to Cart
Add to Cart
Includes
DryEase™ Large Gel Drying Frame (1 frame, 8 clamps), DryEase™ Large Pre-Cut Cellophane (100 sheets), and Gel-Dry™ Drying Solution (500mL).
Accommodate larger format gels or four mini-gels
The Large Gel Drying Kit dries gels using the same components as the DryEase™ Mini-Gel Drying System. Consistently dry gels without cracking and can accommodate up to four mini-gels or a large format gel. Base not required for assembly.
Gel Drying, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage The Large Gel Drying Kit contains a DryEase™ Large Gel Drying Frame (1 frame, 8 clamps), DryEase™ Large Pre-Cut Cellophane (100 sheets), and Gel-Dry™ Drying Solution (500 ml). Store solution at room temperature. Guaranteed stable for one year when properly stored.
Quantity 1 kit
Product Line Novex
Product Type Large Gel Drying Kit
I dried my gel using your DryEase Mini Gel Drying system and the gel cracked during drying. Can you please help?
Please see below for possible causes and solutions:
- Air trapped between cellophane layers. Apply adequate Gel-Drying solution to prevent bubbles between the gel and cellophane assembly.
- Tension on cellophane too strong. Take care not to stretch the cellophane during dryer assembly.
- Rough edges or small cracks on gel edges. Trim rough edges from the gel to eliminate starting points for cracks.
- Cellophane slips during drying. Make sure clamps hold all four edges of the dryer assembly firmly.
I am using your DryEase Mini Gel Drying system and would like to peel off the cellophane after drying the gel but it seems really difficult. Can you please offer some tips?
Usually it is not necessary to peel off the cellophane. If your signal is very faint and you are concerned about getting an exposure, you can rub the top surface of your gel with a damp towel and then peel off the cellophane. Then air dry the gel for a few minutes and perform autoradiography.
I dried my gel using Gel-Drying Solution and it dried milky white. Can the gel be recovered?
Gels turn white when they are dried too rapidly, usually in an environment that is too ventilated, drafty, or warm. You can remedy this problem by dabbing the gel with deionized water on a Kimwipe tissue to rehydrate the white areas. Re-dry the gel in a dark, dry, non-drafty place, such as a drawer or cabinet.
What is the pore size of the DryEase cellophane?
The pore size of DryEase cellophane is 40 microns.
Do you offer a system for drying of protein gels?
We offer the following systems for drying of protein gels:
DryEase Mini Gel Drying System (Cat. No. NI2387): Simple-to-use kit for even, crack-free drying of mini gels that dries gels evenly, using passive evaporation. Gels dried with this system are suitable for fluorography, densitometry, autoradiography, or permanent gel storage. Each DryEase Mini-Gel Drying System contains the following components:
- 2-DryEase Mini-Gel Drying Frames (includes 2 frames + 8 clamps) (Cat. No. NI2380)
- 1-DryEase Mini-Gel Drying Base (Cat. No. NI2300)
- 200-DryEase Mini Cellophane precut sheets (Cat. No. NC2380)
- 500 mL Gel-Dry Drying Solution (Cat. No. NI2100)
Large Gel Drying Kit (Cat. No. NI2207): Provides the same even, crack-free gel drying seen with the DryEase Mini-Gel Drying System but is designed to accommodate larger gels or 4 mini-gels. A base for the system is not required.
Each Large Gel Drying Kit contains the following components:
- 1-DryEase Large Gel Drying Frame (includes 8 clamps) (Cat. No. NI2200)
- 100-DryEase Large Cellophane precut sheets (Cat. No. NC2200)
- 500 mL Gel-Dry Drying Solution (Cat. No. LC4025)
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.