Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Automated Maintenance Plate (Luminex™ System)

Catalog No. CN020601
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CN020601 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CN020601 Supplier Invitrogen™ Supplier No. CN020601
Add to Cart
Add to Cart

Compatible with Luminex 100 and 200 Systems

Automated Maintenance Plate (AMP) allows for easy walk-away maintenance and calibration.

Cell Analysis, Immunoassays

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at RT
Product Line Luminex
Product Type Maintenance Plate
For Use With (Equipment) Luminex Instruments
Quantity 1 Ea.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.