Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ pNPP Solution

Catalog No. 002212
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 ML
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
00-221-2 100 ML
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 00-221-2 Supplier Invitrogen™ Supplier No. 002212
  • $193.00
    $185.65 / Each of 1

    Save $7.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use in immunoassays

May be used as a substrate for alkaline phosphatase (AP) in enzyme immunoassays.

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2-8°C
Detection Method Colorimetric
Product Type pNPP Solution
Substrate pNPP
Substrate Properties Chemical Substrate
Substrate Type AP (Alkaline Phosphatase) Substrate
Quantity 100 ML
Shipping Condition Wet Ice
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.