Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Polypropylene Columns (Empty)

Catalog No. R64050
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R64050 50 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R64050 Supplier Invitrogen™ Supplier No. R64050
Get 16% Off Protein Analysis Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Empty, 10mL columns

Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Format Empty Column
Product Type Polypropylene Column
Content And Storage Store at room temperature.
Quantity 50 Columns

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.