Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Secondary Antibody Solution Alk-Phos. Conjugated (Anti-Mouse)

Catalog No. WP20006
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL (2 Bottles)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WP20006 100 mL (2 Bottles)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WP20006 Supplier Invitrogen™ Supplier No. WP20006
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Pre-diluted, ready-to-use formulation containing alkaline phosphatase labeled polyclonal antibodies in a proprietary stabilizing reagent.

A goat anti-mouse polyclonal alk-phos conjugate, this Secondary antibody is compatable with both chemiluminescent and chromogenic methods of immunodetection, and will identify antigens bound with a primary mouse antibody.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Host Species Goat
Quantity 100 mL (2 Bottles)
Whole Molecule Whole Antibody
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store in refrigerator (2–8°C).
Form Liquid
Isotype IgG
Reactivity Mouse
Shipping Condition Wet Ice
Show More Show Less
Can I purchase the Secondary Antibody solutions from the WesternBreeze Chemiluminescent Detection kits as standalone products?

The goat anti-mouse and goat anti-rabbit secondary antibody solutions AP-conjugated are available as standalone products (Cat. Nos. WP20006 and WP20007), but the rabbit anti-goat secondary antibody solution AP-conjugated is not available as a standalone product.

Can I purchase the Secondary Antibody solutions from the WesternBreeze Chromogenic Detection kits as standalone products?

The goat anti-mouse and goat anti-rabbit secondary antibody solutions AP-conjugated are available as standalone products (Cat. Nos. WP20006 and WP20007) but the rabbit anti-goat secondary antibody solution AP-conjugated is not available as a standalone product.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.