Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Tau (Phospho) [pT231] ELISA Kit

Catalog No. KHB8051
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KHB8051 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KHB8051 Supplier Invitrogen™ Supplier No. KHB8051
Only null left
Add to Cart
Edge
Add to Cart

ELISA

Kit is designed to detect and quantify the level of Tau (Phospho) [pT231] in human cerebrospinal fluid, tissue homogenates, buffered solutions and cell culture supernatants. The assay recognizes both natural and recombinant Tau (Phospho) [pT231]. Principle of the method A monoclonal capture antibody specific for Tau (Phospho) [pT231] has been coated onto the wells of the 96-well plate. During the first incubation, standards of known content and unknown samples are pipetted into the wells and the antigen binds to the immobilized (capture) antibody. After washing, a rabbit antibody specific for the target protein is added to the wells and serves as a detection antibody by binding to the immobilized protein captured during the first incubation. After washing, a horseradish peroxidase labeled anti-rabbit IgG is added. This binds to the detection antibody to complete the four member sandwich. After a third incubation and washing to remove all the unbound enzyme, a substrate solution (TMB) is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of target protein present in the original specimen and the optical density can be read on a standard microplate reader. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10636
Assay Range 1.56 to 100 U/mL
Assay Sensitivity <1 U/mL
Concentration 1.56 to 100 U/μL
Conjugate HRP
Product Type ELISA
Sample Type Cerebrospinal Fluid, Supernatant, Tissue Homogenates
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4137
Gene Symbol DDPAC,FTDP-17 L,MSTD,MTBT1,MTBT2,PPND,PPP1R103,TAU,MAPT
Incubation Time 4 hr.
Interassay CV 7.8%
Intraassay CV 5.4
Kit Contents Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Detection Antibody, anti-rabbit-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Marker Gene Tau
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cerebrospinal Fluid 10 μL, Supernatant 10 μL, Tissue Homogenate 10 μL
Storage Requirements 2°C to 8°C
Target Tau
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.