Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Novex™ Tricine SDS Running Buffer (10X)

Catalog No. LC1675
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC1675 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC1675 Supplier Invitrogen™ Supplier No. LC1675
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Novex Tricine SDS Running Buffer (10X) is formulated for separation of proteins in their denatured state on tricine gels.

Novex Tricine SDS Running Buffer (10X) is formulated for separation of proteins in their denatured state on tricine gels. Tricine gels are specifically designed for the resolution of low molecular weight proteins. In this buffer system, tricine substitutes glycine in the running buffer, resulting in more efficient stacking and destacking of low molecular weight proteins and higher resolution of smaller peptides.

Recommended buffers: Use with tricine sample buffer (Cat. No. LC1676) to obtain optimal separation with tricine SDS running buffer.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sample Loading Buffer
Concentration 10X
Physical Form Liquid
Product Line Novex
Quantity 500 mL
Solubility Information Water
Can urea be used with the precast Tricine gel electrophoresis system to achieve denatured results?

Adding urea to the sample and running buffers, in conjunction with SDS, may provide improved solubilization of the sample if denaturation by SDS does not prove to be sufficient. This must be tested empirically for the protein of interest.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.