Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZOOM™ Thiourea

Catalog No. ZT10002
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ZT10002 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ZT10002 Supplier Invitrogen™ Supplier No. ZT10002
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

The ZOOM™ IEF Fractionator Combo Kit includes the ZOOM™ IEF Fractionator; one pack ZOOM™ Disks of each pH; and ZOOM™ Reagents (urea, thiourea, CHAPS, carrier ampholytes pH 3-10, anode buffers, and cathode buffers).

Offers a fast, reliable method to reduce sample complexity, enrich low abundance proteins, and increase the dynamic range of detection

Solution phase isoelectric focusing with the ZOOM™ IEF Fractionator provides reproducible separations in three hours. Fractionated samples are ready for further analysis by two dimensional gel electrophoresis (2DE), one dimensional gel electrophoresis (1DE), or two dimensional liquid chromatography/mass spectrometry (2D LC/MS).

Sample Chamber and Fractionator Unit Set-up Procedure
The sample chamber and components fit together easily and offer a leak-proof seal. Simply place disks between the sample chambers in the ZOOM™ IEF Fractionator to allow separation in a specific pH range. For example, if you want to fractionate proteins between pH 4.6 and 5.4, flank a sample chamber with a ZOOM™ Disk of pH 4.6 and one of pH 5.4.

Downstream Processing by 2D Gel Electrophoresis
Following fractionation, the separated samples can be subjected to further analysis using narrow or broad range ZOOM™ Strips on the ZOOM™ IPGRunner™ System. Follow with second dimension analysis using neutral pH NuPAGE™ ZOOM™ Gels.

  • ZOOM™ IEF Fractionator is built for safety, reliability, and durability. It includes easy-to-assemble sample chambers, in-built buffer chambers, buffer trough, removable electrode assembly, and safety lid.
  • ZOOM™ Disks are pre-cast polyacrylamide gels that eliminate the need for manual preparation and minimize any chance of cross contamination. These immobilized buffered disks are pre-labeled, disposable, and designed for single use, ensuring consistent and reproducible fractionation. They resolve samples into as many as 7 fractions (using 8 disks of specific pH) to six fractions (using six disks of specific pH) from pH 3-12. Simply place each disk between the sample chambers in the ZOOM™ IEF Fractionator to allow separation in a specific pH range.
  • ZOOM™ Reagents are proteomic grade reagents, including ZOOM™ 2D Protein Solubilizers urea, thiourea, CHAPS, and ampholytes, that ensure optimal conditions for sample preparation.
  • Basic Protein Kit which includes ZOOM™ Discs and ZOOM Strips for IEF of basic proteins, focusing buffers, and solubilizer kits.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Line ZOOM
Product Type ZOOM™ Reagents
Quantity 1 kg
Content And Storage The ZOOM™ IEF Fractionator Combo Kit includes the ZOOM™ IEF Fractionator; one pack ZOOM™ Disks of each pH; and ZOOM™ Reagents (urea, thiourea, CHAPS, carrier ampholytes pH 3-10, anode buffers, and cathode buffers). ZOOM™ Disks, Carrier Ampholytes, and Buffers are stored at +4°C.
Format Powder
Shipping Condition Room Temperature
Form Powder
Matrix IEF Media
What are the components of the ZOOM IEF Fractionator?

The ZOOM IEF Fractionator includes the following components:

Chamber Assembly Tube with Anode Reservoir
Spill Trough with Cathode Reservoir Lid
Sample Chambers (7)
Sample Chamber O-ring Seals, red (10)
Sample Chamber Caps with O-rings (7)
Cathode End Sealer
Anode End Sealer
Cathode End Screw Cap
Spacers, black (8)
Spares Box 1
Sample Chamber O-ring Seals (8)
Sample Chamber Caps with O-rings (7)
Spares Box 2
Cathode Chamber Seals (2)
Spacers, black (8)

Can I heat my protein sample prepared using ZOOM 2D Protein Solubilizer?

We do not recommend heating protein samples containing urea over 37 degrees C as elevated temperatures cause urea to hydrolyze to isocyanate, which modifies proteins by carbamylation.

How do you recommend storing protein samples prepared in the Sample Rehydration buffer?

We recommend storing them at -80 degrees C. We do not recommend storing them at -20 degrees C

How much ampholyte do you recommend adding to the sample rehydration buffer?

The recommended ampholyte concentration in the sample rehydration buffer is 0.5%.

*If you are loading 5-50 µg of protein (pure protein or crude lysate) per ZOOM strip, use 0.5% ampholytes in the sample rehydration buffer.
*If you are loading >50 µg of protein (crude lysate or fractionated sample) per ZOOM Strip, use 0.5-2% ampholytes in the sample rehydration buffer.

Note: Higher ampholyte concentration requires longer focusing times.

What is the maximum recommended volume of protein sample to be added to the Sample Rehydration buffer?

The maximum volume of the protein sample should at most be 1/6 of the final sample volume that will be added to the strip. A good general volume would be 5-10µL. 140 µL of sample diluted in Sample Rehydration buffer is used to rehydrate each ZOOM Strip for the standard rehydration time of one hour. For overnight rehydration, we recommend using 155 µL of diluted sample.

What is the purpose of alkylation prior to IEF?

Alkylation prevents unwanted protein modifications by alkylating cysteines to avoid mixed disulfide formation and reoxidation and this allows for crisper focusing.

Do you still offer ZOOM 2D Protein Solubilizers?

Sorry we have discontinued selling the ZOOM 2D Protein Solubilizers.

What are the main reasons for performing 2D gel electrophoresis of proteins?

The main advantages of performing 2D gel electrophoresis of proteins and applications used for are listed below:

Advantages:

*Simultaneous separation of hundreds to thousands of proteins
*High capacity with superior resolution
*Compatible with further analysis by MS for protein identification and sequencing
Ability to separate and analyze low-abundance proteins

Applications:

*Comparative proteomics: identifying and analyzing differences between complex mixtures of proteins
*Protein profiling, biomarker discovery
*Separation and analysis of protein variants and isoforms

What products do you offer for 2D gel electrophoresis of proteins?

We offer the following products for the first- and second-dimension separation of proteins:

First-dimension separation:

*Vertical gels for separation of proteins based on their isoelectric point (pI) (https://www.thermofisher.com/us/en/home/life-science/protein-expression-and-analysis/protein-gel-electrophoresis/protein-gels/specialized-protein-separation/isoelectric-focusing.html)

*Solution-phase isoelectric focusing of proteins using ZOOM Disks (immobilized buffer disks of specific pH) to reduce sample complexity, enrich low-abundance proteins, and increase the dynamic range of detection (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/specialized-protein-gels/isoelectric-focusing/zoom-ief-fractionator.html)

*Mini gel system for high-throughput isoelectric focusing of proteins using ZOOM IPG (Immobilized pH Gradient) Strips (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/specialized-protein-gels/2d-gel-electrophoresis/zoom-ipgrunner-system.html)

Second-dimension separation:

*ZOOM gels for 2D electrophoresis: NuPAGE Bis-Tris (Cat. No. NP0330BOX) and Tris-Glycine (Cat. No. EC60261BOX) mini gels with IPG wells ( to accommodate 7 cm ZOOM strips) for separation of proteins based on their molecular weight

What is 2D protein gel electrophoresis?

2D protein gel electrophoresis is the separation of proteins in two dimensions. In the first dimension, proteins are separated by their isoelectric point (pI) using isoelectric focusing, and in the second dimension, they are separated by their mass using SDS-PAGE.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.