Learn More
Crescent Chemical Co Inc NOVOBIOCIN-NA-SALT RESEARCH
SDP
Catalog No. 50247951
Encompass_Preferred
Novobiocin-Na-salt research grade Coumarin-glycoside antibiotic. Inhibitor of bacterial DNA gyrase (1). Mechanism of action (2). Antagonist of heat shock protein 90 (Hsp90) (3, 4, 5). Size - 1G Storage Conditions - +2 °C TO +8 °C Catalog Number - 30995
Catalog No. 50-247-951 Supplier Crescent Chemical Co Inc Supplier No. SE30995.01
Novobiocin-Na-salt research grade Coumarin-glycoside antibiotic. Inhibitor of bacterial DNA gyrase (1). Mechanism of action (2). Antagonist of heat shock protein 90 (Hsp90) (3, 4, 5). Size - 1G Storage Conditions - +2 °C TO +8 °C Catalog Number - 30995