Learn More
Selleck Chemical LLC Novobiocin Sodium (NSC 2382) S2492-50mg
SDP
Catalog No. 503111749
Encompass_Preferred
Novobiocin Sodium (NSC 2382 Albamycin Cathomycin) is an aminocoumarin antibiotic that targets bacterial DNA gyrase (TopoIV) used to treat susceptible gram positive bacteria
Catalog No. 50-311-1749 Supplier Selleck Chemical LLC Supplier No. S249250MG
Add to Cart
Add to Cart
Novobiocin Sodium (NSC 2382 Albamycin Cathomycin) is an aminocoumarin antibiotic that targets bacterial DNA gyrase (TopoIV) used to treat susceptible gram positive bacteria