Learn More
Alkali Scientific Novobiocin Sodium Salt, 25 G
SDP
Catalog No. 5000661322
Encompass_Preferred
Novobiocin Sodium Salt is an antibiotic used for bacterial inhibition in microbiological applications. This 25g powder form is highly effective against Gram-positive bacteria, inhibiting their growth by interfering with bacterial DNA replication. It is commonly used in laboratory settings for the selective inhibition of specific bacterial species, particularly for research involving Staphylococcus species and other Gram-positive microorganisms. Novobiocin is used in various molecular biology and bacteriological tests, including susceptibility testing, and can be incorporated into culture media or used as a standalone antimicrobial agent. The powder form ensures easy handling and accurate preparation of solutions for experimental use.
Catalog No. 50-006-61322 Supplier Alkali Scientific Supplier No. NOV003CP
Novobiocin Sodium Salt is an antibiotic used for bacterial inhibition in microbiological applications. This 25g powder form is highly effective against Gram-positive bacteria, inhibiting their growth by interfering with bacterial DNA replication. It is commonly used in laboratory settings for the selective inhibition of specific bacterial species, particularly for research involving Staphylococcus species and other Gram-positive microorganisms. Novobiocin is used in various molecular biology and bacteriological tests, including susceptibility testing, and can be incorporated into culture media or used as a standalone antimicrobial agent. The powder form ensures easy handling and accurate preparation of solutions for experimental use.