Learn More
Alkali Scientific Novobiocin Sodium Salt, 5 G
SDP
Catalog No. 5000660900
Encompass_Preferred
Novobiocin Sodium Salt is a powerful antibiotic formulated in a 5g powder form, effective in inhibiting Gram-positive bacteria. By interfering with bacterial DNA synthesis, it prevents bacterial replication and is commonly used in microbiology for selective bacterial inhibition. This antibiotic is particularly useful in isolating and identifying Staphylococcus species, often applied in research and clinical settings. Novobiocin is also widely used in laboratory experiments involving antimicrobial resistance studies. The powder format allows easy preparation and precise dosage for media supplementation or solution preparation in antimicrobial susceptibility testing.
Catalog No. 50-006-60900 Supplier Alkali Scientific Supplier No. NOV004CP
Novobiocin Sodium Salt is a powerful antibiotic formulated in a 5g powder form, effective in inhibiting Gram-positive bacteria. By interfering with bacterial DNA synthesis, it prevents bacterial replication and is commonly used in microbiology for selective bacterial inhibition. This antibiotic is particularly useful in isolating and identifying Staphylococcus species, often applied in research and clinical settings. Novobiocin is also widely used in laboratory experiments involving antimicrobial resistance studies. The powder format allows easy preparation and precise dosage for media supplementation or solution preparation in antimicrobial susceptibility testing.