Learn More
Enzo Life Sciences Novobiocin . sodium salt (5g). CAS: 1476-53-5
SDP
Supplier:  Enzo Life Sciences BMLA2560005
Encompass_Preferred
Antibiotic. Inhibitor of HSP90. Interacts with the C-terminal ATP-binding domain of HSP90 in contrast to the benzoquinone ansamycins 17-AAG (BML-EI308), geldanamycin (BML-EI280), herbimycin A (BML-EI227) and the chemically unrelated radicicol (BML-EI285), which bind to the N-terminal ATP-binding site of HSP90. Inhibitor of DNA gyrase. Useful for the production of positively supercoiled plasma DNA, targeting the nucleotide-binding site of gyrase B. Inhibits retrovirus RNA-dependent DNA polymerase. Potent inhibitor of ADP ribosylation. Inhibits LPS-induced production of pro-inflammatory cytokines, such as TNF-α, IL-1, IL-6 and IL-10. Alternative name: Albamycin, Streptonivicin, N-[7-[[3-O-Aminocarbonyl-6-deoxy-5-C-methyl-4-O-beta-L-lyxo-hexapyranosyl]oxy]-4-hydroxy-8-methyl-2-oxo-2H-1-benzopyran-3-yl]-4-hydroxy-3-(3-methyl-2-butenyl)benzamide. Solubility: Soluble in water (100mg/ml), ethanol or DMSO. Long Term Storage: +4°C.
Catalog No. 50-201-0236