Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novobiocin Sodium Salt, G-Biosciences
SCI_ED

Catalog No. S157218
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157218 10 g
S000024 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157218 Supplier G-Biosciences Supplier No. RC1072
Add to Cart
Add to Cart

Novobiocin is used to study heat shock protein inhibition 1 and to produce positively supercoiled plasmid DNA.

It is an inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase. It also is an inhibitor of retrovirus RNA-dependent DNA-polymerase.

Chemical Identifiers

Molecular Weight (g/mol) 634.6 g/mol

Specifications

Boiling Point 848.2°C
Quantity 10 g
Percent Purity >95%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.