Learn More
Selleck Chemical LLC Novobiocin Sodium10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S249210MM/1ML
Encompass_Preferred
Novobiocin Sodium (NSC 2382, Albamycin, Cathomycin) is an aminocoumarin antibiotic that targets bacterial DNA gyrase (TopoIV), used to treat susceptible gram positive bacteria. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3086