Learn More
TOKU-E NOVOBIOCIN SOLN 10X1ML
Catalog No. NC2068771
Encompass_Preferred
Antibiotic,ReadyMade™, sterile-filtered, 100 mg/ml in water. Useful in cancer research, cell culture, susceptibility testing.
Catalog No. NC2068771 Supplier TOKU-E Supplier No. N08910X1ML
Antibiotic,ReadyMade™, sterile-filtered, 100 mg/ml in water. Useful in cancer research, cell culture, susceptibility testing.