Learn More
TOKU-E Co-trimoxazole ReadyMade™ Solution | CAS 8-64-90-2 | 100 mg/ML in DMSO | sterile-filtered | 10 x 1 ML
Catalog No. NC2068765
Encompass_Preferred
Co-trimoxazole ReadyMade™ Solution | CAS 8-64-90-2 | 100 mg/ML in DMSO | sterile-filtered | 10 x 1 ML
Catalog No. NC2068765 Supplier TOKU-E Supplier No. N0891ML
Co-trimoxazole ReadyMade™ Solution | CAS 8-64-90-2 | 100 mg/ML in DMSO | sterile-filtered | 10 x 1 ML