Learn More
Agilent Technologies NOVOCLEAN SOLUTION 1X 500 ML
Catalog No. NC2388524
Encompass
  • Ready-to-use liquid cleaning reagent
  • 1X concentrated solution
  • Volume: 500 mL
  • Effectively removes organic residues and contaminants
  • Reduces build-up on critical components
  • Compatible with Agilent LC and LC/MS systems
  • Enhances system longevity and reliability
  • Improves analytical accuracy
  • Minimizes instrument downtime
  • Reduces sample carryover and cross-contamination
  • Suitable for pharmaceutical analysis, proteomics, and environmental testing
  • Country of origin: Germany
  • Net weight: 0.500 kg
  • Gross weight: 0.600 kg
Catalog No. NC2388524 Supplier Agilent Technologies Supplier No. 872B622
  • Ready-to-use liquid cleaning reagent
  • 1X concentrated solution
  • Volume: 500 mL
  • Effectively removes organic residues and contaminants
  • Reduces build-up on critical components
  • Compatible with Agilent LC and LC/MS systems
  • Enhances system longevity and reliability
  • Improves analytical accuracy
  • Minimizes instrument downtime
  • Reduces sample carryover and cross-contamination
  • Suitable for pharmaceutical analysis, proteomics, and environmental testing
  • Country of origin: Germany
  • Net weight: 0.500 kg
  • Gross weight: 0.600 kg