Learn More
Abcam Novokinin, Angiotensin AT2 receptor agonist, 1MG
Catalog No. 502705439
Encompass_Preferred
MW 795.98 g/mol, Purity >98%. Angiotensin AT₂ receptor agonist (Ki = 7 μM). Antagonises the antinociceptive effect of morphine, shows antihypertensive, vasorelaxing and anorexigenic effects in vivo. Orally active. Shows central effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5439 Supplier Abcam Supplier No. AB1450581MG
MW 795.98 g/mol, Purity >98%. Angiotensin AT₂ receptor agonist (Ki = 7 μM). Antagonises the antinociceptive effect of morphine, shows antihypertensive, vasorelaxing and anorexigenic effects in vivo. Orally active. Shows central effects.
The product is subject to the following: Abcam Restricted Use Statement